missing translation for 'onlineSavingsMsg'
Learn More

TIN2 Antibody (5G11), Novus Biologicals™

Product Code. 18359341 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359341 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359341 Supplier Novus Biologicals Supplier No. NBP315221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TIN2 Monoclonal antibody specifically detects TIN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIN2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5G11
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias (TRF1)-interacting nuclear factor 2 variant 1, TERF1 (TRF1)-interacting nuclear factor 2, TERF1-interacting nuclear factor 2, TIN2TRF1-interacting nuclear protein 2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIN2. UniProt ID (Q9BSI4).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26277
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.