missing translation for 'onlineSavingsMsg'
Learn More

Tin wire, 1.0mm (0.04in) dia, Puratronic™, 99.998% (metals basis)

Product Code. 11376358
Change view
Click to view available options
Quantity:
50 cm
250 cm
1000 cm
Unit Size:
1000cm
250cm
50cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376358 250 cm 250cm
11366358 50 cm 50cm
11386358 1000 cm 1000cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11376358 Supplier Thermo Scientific Alfa Aesar Supplier No. 011905.CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 5.72g/m | CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver to Gray
Appearance Silver Gray wire
Quantity 250 cm
Odor Odorless
Assay Percent Range 99.998% (Metals basis)
Form Wire
Chemical Name or Material Tin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.