missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Tin Cell Test Kit

Product Code. 11784785
Change view
Click to view available options
Range:
0.10 - 2.50mg/I Sn
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
11784785 0.10 - 2.50mg/I Sn Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11784785 Supplier WTW™ Supplier No. 250401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

With certificate for optimum precision.

  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For infrequent or very large sample throughput in the laboratory
  • Sample adding, inserting, measuring and reading
  • Reaction cuvette with barcode or method selection

TRUSTED_SUSTAINABILITY

Specifications

Analytes Pyrocatechol violet
For Use With (Application) For waste water, electroplating bath solutions
For Use With (Equipment) PhotoLab™ S12, PhotoLab™ Spectral photometers
Includes Certificates
No. of Tests 25
Range 0.10 - 2.50mg/I Sn
Sufficient For 25 Tests
Test Method Photometric
Type Tin (Sn) cell test kit
compliance-icons
Product Identifier
  • tin cell test method
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid, N-cetyl-N,N,N-trimethylammonium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.