missing translation for 'onlineSavingsMsg'
Learn More

Tin Standard 1000mg/L in 20% HCL (Assurance Grade), SPEX CertiPrep

Product Code. 10204152 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
PTFE
Triple-leached LDPE bottle
Unit Size:
250mL
30mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204152 250 mL PTFE 250mL
16321256 30 mL Triple-leached LDPE bottle 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10204152 Supplier SPEX Certiprep Supplier No. PLSN52T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Tin Standard
  • Made with ASTM Type I Water
  • 1,000 μg/mL concentration
  • 250 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Chemical Identifiers

CAS 7440-31-5 , 7647-01-0 , 7732-18-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Packaging PTFE
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS Min % 1.0, 20.0, 79.0
CAS Max % 1.0, 20.0, 79.0
Technique AA, ICP
Type Certified Reference Material
Quantity 250 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
1% HNO3/1% HF    1000μg/mL    PLSN2-2X
2% HNO3/2% HF    10,000    PLSN2-3X
20% HCl    1000μg/mL    PLSN5-2Y, -2T, -2X    10,000    PLSN5-3Y, -3X
Chemical Name or Material Tin Standard
compliance-icons compliance-icons
Product Identifier
  • 1000
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P364-And wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.