missing translation for 'onlineSavingsMsg'
Learn More

Tin slug, 11mm (0.43 in.) dia. x 2mm (0.1 in.) thick, Puratronic™, 99.995% (metals basis), Thermo Scientific™

Product Code. 11460351
Change view
Click to view available options
Quantity:
10 Pc.
50 Pc.
Unit Size:
Pack of 10
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460351 10 Pc. Pack of 10
11369547 50 Pc. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460351 Supplier Thermo Scientific Alfa Aesar Supplier No. 041169.KK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Evaporation source

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 10 Pc.
Odor Odorless
Weight ≈1.3g
Form Slug
Percent Purity 99.995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Tin slug, 11mm (0.43 in.) dia. x 2mm (0.1 in.) thick
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.