missing translation for 'onlineSavingsMsg'
Learn More

Tin shot, 3mm (0.1in), 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11391618
Click to view available options
Quantity:
25 g
100 g
500 g
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray shot
Assay Percent Range 99.999% (Metals basis)
Quantity 25 g
Odor Odorless
Form Shot
Chemical Name or Material Tin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'