missing translation for 'onlineSavingsMsg'
Learn More

Tin selenide, 99.999% (metals basis)

Product Code. 11353477
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353477 25 g 25g
11363477 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353477 Supplier Thermo Scientific Alfa Aesar Supplier No. 018781.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1315-06-6 | H2SeSn | 199.70 g/mol

Tin selenide finds application in low- cost photovoltaics devices and memory-switching devices. It acts as a solid-state lubricant. It is also used for solar cells and anodes for lithium-ion batteries. Further, it is a narrow band-gap semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-06-6
Molecular Formula SnSe
Molecular Weight (g/mol) 197.65
MDL Number MFCD00054049
InChI Key MFIWAIVSOUGHLI-UHFFFAOYSA-N
IUPAC Name selenoxo-lambda~2~-stannane
SMILES [Se]=[SnH2]

Specifications

Melting Point 861°C
Physical Form Pieces
Quantity 25 g
Assay Percent Range 99.999% (metals basis)
Solubility Information Insoluble in water. Soluble in aqua regia,alkali sulfides,and alkali selenides.
Formula Weight 197.65
Chemical Name or Material Tin selenide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.