missing translation for 'onlineSavingsMsg'
Learn More

Tin powder, spherical, APS approximately 0.1 micron, Thermo Scientific Chemicals

Catalog Number 11375777
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]
This item is not returnable. View return policy
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3089
Quantity 25 g
Odor Odorless
Particle Size APS ≈ 0.1 Micron
Form Spherical Powder
Chemical Name or Material Tin powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'