missing translation for 'onlineSavingsMsg'
Learn More

Tin powder, -325 mesh, Thermo Scientific Chemicals

Catalog Number 11325838
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]
This item is not returnable. View return policy
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
5352426
CHEBI:32990
tin
[Sn]

Apparent density 3.2g/cm3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray powder
Assay Percent Range 99.8% (Metals basis)
Quantity 100 g
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Tin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'