missing translation for 'onlineSavingsMsg'
Learn More

Tin oxide, antimony doped, polymeric precursor, Oxide ≈ 30 wt%, Thermo Scientific Chemicals

Catalog Number 11354727
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1332-29-2
MFCD00011244
Antimony doped tin oxide
This item is not returnable. View return policy
1332-29-2
MFCD00011244
Antimony doped tin oxide

Used in transparent electrical conducting electrodes

Used in transparent electrical conducting electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Viscous Liquid
Quantity 25 g
Solubility Information Soluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈30 wt%
Density 1.61 g/mL
Chemical Name or Material Tin oxide, Antimony doped, Polymeric precursor

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'