missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) phthalocyanine dichloride

Product Code. p-7050867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354537 1 g 1g
11364537 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354537 Supplier Thermo Scientific Alfa Aesar Supplier No. 039606.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18253-54-8 | C32H16Cl2N8Sn | 702.146 g/mol

Tin(IV) phthalocyanine dichloride is a stable metal phthalocyanine dye utilized in biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18253-54-8
Molecular Formula C32H16Cl2N8Sn
Molecular Weight (g/mol) 702.146
MDL Number MFCD00054210
InChI Key BBRNKSXHHJRNHK-UHFFFAOYSA-L
Synonym tin iv phthalocyanine dichloride, sncl2pc, dichlorotin phthalocyanine, tin phthalocyanine dichloride, tin iv phthalocyaninedichloride
PubChem CID 9580361
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C6N4[Sn](N7C(=NC2=N3)C8=CC=CC=C8C7=NC9=NC(=N6)C1=CC=CC=C19)(Cl)Cl

Specifications

Quantity 1 g
UN Number UN3146
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 702.34
Physical Form Powder
Chemical Name or Material Tin(IV) phthalocyanine dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.