missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) phthalocyanine dichloride, Thermo Scientific Chemicals
44.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
Tin(IV) phthalocyanine dichloride is a stable metal phthalocyanine dye utilized in biochemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 g |
UN Number | UN3146 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 702.34 |
Physical Form | Powder |
Chemical Name or Material | Tin(IV) phthalocyanine dichloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'