missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Color | White to Gray |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.9% |
Linear Formula | SnO2 |
Merck Index | 15, 8905 |
Solubility Information | Solubility in water: insoluble. Other solubilities: slowly soluble in sodium hydroxide solution, insoluble in alcohol and cold acids |
Formula Weight | 150.69 |
Percent Purity | 99.9% |
Grade | Trace Metal Basis |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'