missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) isopropoxide isopropanol adduct, 98% (metals basis)

Product Code. 11389647
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389647 5 g 5g
11399647 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389647 Supplier Thermo Scientific Alfa Aesar Supplier No. 041269.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 132951-93-0

Tin(IV) isopropoxide isopropanol adduct is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, electrochemical and conducting properties and thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132951-93-0
MDL Number MFCD00145408

Specifications

Melting Point ∼130°C (decomposition)
Boiling Point 131°C (1.6 mmHg)
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in hydrocarbons and warm isopropanol. Hydrolyzes with water. Reacts with alcohols,ketones and esters.
Formula Weight 414.15
Percent Purity 98%
Physical Form Wet Solid
Assay (metals basis)
Chemical Name or Material Tin(IV) isopropoxide isopropanol adduct

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.