missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) isopropoxide, 98% (metals basis), 10% w/v in isopropanol/toluene

Product Code. 11367988
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367988 100 mL 100mL
11377988 5 mL 5mL
11387988 25 mL 25mL
11475180 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11367988 Supplier Thermo Scientific Alfa Aesar Supplier No. 036563.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1184-61-8 | C12H32O4Sn | 359.094 g/mol

Tin(IV) isopropoxide isopropanol is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, thin films doped tin oxide nanoparticles, thin films and are widely studied as gas sensors. Tin(IV) precursor solution was prepared from tin(IV) isopropoxide dissolved in isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-61-8
Molecular Formula C12H32O4Sn
Molecular Weight (g/mol) 359.094
MDL Number MFCD00145408
InChI Key VUDAJANAMZGVOP-UHFFFAOYSA-N
Synonym tin iv isopropoxide, tin tetraisopropoxide, acmc-20aji2, tin 4+ tetrapropan-2-olate, tin 4+ ion tetrakis propan-2-olate, tin 4+ tetrakis propan-2-olate
PubChem CID 85354423
IUPAC Name propan-2-ol;tin
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Sn]

Specifications

Quantity 100 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Soluble in hydrocarbons,reacts with alcohols,ketones and esters
Formula Weight 355.06
Percent Purity 98%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tin(IV) isopropoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.