missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) iodide, ultra dry, 99.998% (metals basis)

Product Code. 11356878
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356878 25 g 25g
11366878 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356878 Supplier Thermo Scientific Alfa Aesar Supplier No. 013617.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-47-8 | H4I4Sn | 630.36 g/mol

Tin(IV) iodide is used in the preparation of tin(IV) oxide and tin(II) oxide. It reacts with hydroiodic acid to get rare tin hexaiodide. It forms tetraiodobis triphenylphosphine tin complexes by reacting with triphenylphosphine, which finds application in toothpaste formulation. Further, it acts as analytical reagents and also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-47-8
Molecular Formula H4I4Sn
Molecular Weight (g/mol) 630.36
MDL Number MFCD00036272
InChI Key ZBCVLXKKLMNLGV-UHFFFAOYSA-N
Synonym stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide
PubChem CID 24631
IUPAC Name tetraiodostannane
SMILES [Sn].I.I.I.I

Specifications

Boiling Point 340°C
Melting Point ∼143°C
Physical Form Powder
Quantity 25 g
UN Number UN3260
Merck Index 14,8777
Solubility Information Soluble in alcohol,benzene,carbon disulfide,chloroform and ether.
Formula Weight 626.31
Percent Purity 99.998%
Packaging Ampule
Refractive Index 2.106
Sensitivity Moisture sensitive
Density 4.46 g/mL
Assay (metals basis)
Chemical Name or Material Tin(IV) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.