missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) iodide, 95%

Product Code. 11334809
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334809 50 g 50g
11324809 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334809 Supplier Thermo Scientific Alfa Aesar Supplier No. 071114.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-47-8 | H4I4Sn | 630.36 g/mol

Used in chemical research and as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-47-8
Molecular Formula H4I4Sn
Molecular Weight (g/mol) 630.36
MDL Number MFCD00036272
InChI Key ZBCVLXKKLMNLGV-UHFFFAOYSA-N
Synonym stannic iodide, stannane, tetraiodo, tin iv iodide, tetraiodotin, tin tetraiodide, tin iodide, unii-8h688bw69x, tin iv iodide 1:4, tin-iv-iodide
PubChem CID 24631
IUPAC Name tetraiodostannane
SMILES [Sn].I.I.I.I

Specifications

Boiling Point 340°C
Melting Point ∼143°C
Color Red
Physical Form Crystalline
Quantity 50 g
Linear Formula SnI4
UN Number UN3260
Merck Index 14,8777
Solubility Information Soluble in alcohol,benzene,CS2,chloroform,and ether; Decomposes in water
Formula Weight 626.31
Percent Purity 95%
Refractive Index 2.106
Sensitivity Moisture sensitive
Density 4.46 g/mL
Chemical Name or Material Tin (IV) iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.