missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) chloride bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is used as a raw material in synthesis. It is also used as NMR shift reagents and as catalysts for organic synthesis, and precursors to industrial hydroformylation catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 199°C |
Physical Form | Crystalline |
Quantity | 50 g |
UN Number | UN3146 |
Solubility Information | Soluble in organic solvents. |
IUPAC Name | tin(4+) bis(2,4-dioxopentan-3-ide) dichloride |
Formula Weight | 387.83 |
Percent Purity | 95% |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Tin(IV) chloride bis(2,4-pentanedionate) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'