missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride, 99.99% (metals basis)

Product Code. p-4407998
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11314879

Brand: Thermo Scientific Alfa Aesar 071203.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7646-78-8 | Cl4Sn | 260.51 g/mol

Tin(IV) chloride is a precursor to prepare organotin compounds such as tetralkyltin and dialkyldichlorotin(IV), which finds application as catalysts and polymer stabilizers. As a Lewis acid catalyst, it is used in Fridel-Crafts reactions for alkylation and cyclization. It is involved in the selective nitration of aromatic compounds in presence of fuming nitric acid. Furthermore, it is used to prepare tin(IV) oxide coating by sol-gel process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-78-8
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.51
MDL Number MFCD00011242
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl

Specifications

Boiling Point 114°C
Melting Point °C to 33°C
Physical Form Liquid
Quantity 25 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Miscible with alcohol,benzene,toluene,chloroform,acetone,carbon terachloride,gasoline and carbon disulfide.
Formula Weight 260.5
Refractive Index 1.512
Sensitivity Moisture sensitive
Density 2.26 g/mL
Chemical Name or Material Tin(IV) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.