missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(IV) bromide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
A specialty product for proteomics research. Used as a precursor for SnO2 through laser-assisted CVD with higher efficiency than other precursors such as dibutyl tin diacetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 202°C (734mmHg) |
Melting Point | 31°C |
Color | Colorless to White |
Physical Form | Lump |
Quantity | 50 g |
UN Number | UN3260 |
Merck Index | 14,8773 |
Solubility Information | Very soluble in water. Soluble in ethyl alcohol. |
Formula Weight | 438.33 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'