missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) bromide, 99%

Product Code. p-7080309
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364809 50 g 50g
11354809 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364809 Supplier Thermo Scientific Alfa Aesar Supplier No. 071120.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-67-5 | Br4Sn | 438.326 g/mol

A specialty product for proteomics research. Used as a precursor for SnO2 through laser-assisted CVD with higher efficiency than other precursors such as dibutyl tin diacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-67-5
Molecular Formula Br4Sn
Molecular Weight (g/mol) 438.326
MDL Number MFCD00011240
InChI Key LTSUHJWLSNQKIP-UHFFFAOYSA-J
Synonym tin iv bromide, stannic bromide, tin tetrabromide, stannane, tetrabromo, tin perbromide, snbr4, tin bromide, tin iv bromide 1:4, tetrabromotin
PubChem CID 24616
IUPAC Name tetrabromostannane
SMILES Br[Sn](Br)(Br)Br

Specifications

Boiling Point 202°C (734mmHg)
Melting Point 31°C
Color Colorless to White
Physical Form Lump
Quantity 50 g
UN Number UN3260
Merck Index 14,8773
Solubility Information Very soluble in water. Soluble in ethyl alcohol.
Formula Weight 438.33
Percent Purity 99%
Odor Acidic
Sensitivity Air and moisture sensitive
Density 3.34 g/mL
Chemical Name or Material Tin (IV) bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.