missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) 2-ethylhexanoate diisopropoxide, 98% (metals basis), Thermo Scientific Chemicals

Catalog Number 11393538
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
288374-32-3
MFCD00210604
tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis
This item is not returnable. View return policy
288374-32-3
MFCD00210604
tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 523.08
Percent Purity 98%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tin(IV) 2-ethylhexanoate diisopropoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'