missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) phthalocyanine, Thermo Scientific Chemicals

Product Code. 11374537
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374537 2 g 2g
11384537 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374537 Supplier Thermo Scientific Alfa Aesar Supplier No. 039607.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tin(II) phthalocyanine is used as an organic light-emitting diode (OLED) material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15304-57-1
Molecular Formula C32H16N8Sn
Molecular Weight (g/mol) 631.25
MDL Number MFCD00058867
InChI Key LLVONELOQJAYBZ-UHFFFAOYSA-N
Synonym tin ii phthalocyanine, tin phthalocyanine, snpc, phthalocyanatotin cento

Specifications

Quantity 2 g
UN Number UN3146
Formula Weight 631.43
Physical Form Powder
Chemical Name or Material Tin(II) phthalocyanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.