missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(II) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1080°C (decomposition) |
Color | Black-Blue |
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | SnO |
Merck Index | 14,8787 |
Solubility Information | Insoluble in water,and alcohol. |
Formula Weight | 134.69 |
Percent Purity | 99.9% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'