missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxalate, Thermo Scientific Chemicals

Catalog Number 11397707
15.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
814-94-8
C2O4Sn
206.728
MFCD00040678
OQBLGYCUQGDOOR-UHFFFAOYSA-L
stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
13149
oxalate;tin(2+)
C(=O)(C(=O)[O-])[O-].[Sn+2]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
814-94-8
C2O4Sn
206.728
MFCD00040678
OQBLGYCUQGDOOR-UHFFFAOYSA-L
stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
13149
oxalate;tin(2+)
C(=O)(C(=O)[O-])[O-].[Sn+2]

Tin(II) oxalate is used for dyeing and printing textiles. It acts as a catalyst for the esterification reactions between long chain fatty acids and alcohols and in polymerization reactions. It is also used in stannous oral care compositions. Further, it is used in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 280°C (decomposition)
Color White
Density 3.56 g/mL
Linear Formula SnC2O4
Quantity 100 g
UN Number UN3288
Beilstein 3708588
Merck Index 14,8786
Solubility Information Soluble in dilute HCl; Insoluble in water;Soluble in acids; Insoluble in water and acetone.
Formula Weight 206.72
Physical Form Powder
Chemical Name or Material Tin(II) oxalate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'