missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) methoxide

Product Code. 11367977
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367977 5 g 5g
11357977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367977 Supplier Thermo Scientific Alfa Aesar Supplier No. 020897.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14794-99-1 | C2H6O2Sn | 180.778 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14794-99-1
Molecular Formula C2H6O2Sn
Molecular Weight (g/mol) 180.778
MDL Number MFCD00015612
InChI Key UISUQHKSYTZXSF-UHFFFAOYSA-N
Synonym dimethoxytin ii, tin ii methoxide, acmc-1bxxn, methanol, tin 2+ salt 8ci,9ci
PubChem CID 71317297
IUPAC Name methanolate;tin(2+)
SMILES C[O-].C[O-].[Sn+2]

Specifications

Melting Point 242°C to 243°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 180.76
Physical Form Powder
Chemical Name or Material Tin(II) methoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.