missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) methanesulfonate, 50% w/w aq. soln.

Product Code. p-7040118
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
This item is not returnable. View return policy

Product Code. 15466178

Brand: Thermo Scientific Alfa Aesar H59773.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53408-94-9 | C2H6O6S2Sn | 308.89 g/mol

It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53408-94-9
Molecular Formula C2H6O6S2Sn
Molecular Weight (g/mol) 308.89
MDL Number MFCD00137737
InChI Key JALQQBGHJJURDQ-UHFFFAOYSA-L
Synonym stannous methanesulfonate, methanesulfonic acid, tin 2+ salt, tin ii methanesulfonate, methanesulfonate; tin 2+, methanesulfonic acid, tin 2+ salt 2:1, tin ii methanesulfonate solution, tin ii bis methanesulfonate, pubchem11154, stannous methylsulfonate, 2ch3o3s.sn
PubChem CID 6452824
IUPAC Name methanesulfonate;tin(2+)
SMILES [Sn++].CS([O-])(=O)=O.CS([O-])(=O)=O

Specifications

Melting Point -27°C
Density 1.55
Linear Formula (CH3SO3)2Sn
Refractive Index 1.444
Quantity 100 mL
UN Number UN3265
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 308.91
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Chemical Name or Material Tin(II) methanesulfonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.