missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) fluorophosphate, Thermo Scientific Chemicals

Product Code. 11394537
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394537 25 g 25g
11304547 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394537 Supplier Thermo Scientific Alfa Aesar Supplier No. 039608.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52262-58-5
Molecular Formula FO3PSn
Molecular Weight (g/mol) 216.679
MDL Number MFCD00058794
InChI Key ZIQLCYCKKROENE-UHFFFAOYSA-L
Synonym tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+
PubChem CID 44717639
IUPAC Name fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+)
SMILES [O-]P(=O)([O-])F.[Sn+2]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Solubility Information Slightly soluble (3.0g/L at 25°C)
Formula Weight 216.66
Chemical Name or Material Tin(II) fluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.