Learn More
Tin(II) fluoride, 97.5%
Beskrivning
Tin(II) fluoride is used in the prevention of dental caries. It is an active ingredient in toothpastes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Boiling Point | 850°C |
| Melting Point | 219°C |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Linear Formula | SnF2 |
| UN Number | UN3260 |
| Merck Index | 14,8784 |
| Solubility Information | Soluble in potassium hydroxide and potassium fluoride. Insoluble in water,ethanol,ether and chloroform. |
| Formula Weight | 156.69 |
| Visa mer |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.