Learn More
Tin(II) fluoride, 97.5%
Description
Tin(II) fluoride is used in the prevention of dental caries. It is an active ingredient in toothpastes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 850°C |
| Melting Point | 219°C |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Linear Formula | SnF2 |
| UN Number | UN3260 |
| Merck Index | 14,8784 |
| Solubility Information | Soluble in potassium hydroxide and potassium fluoride. Insoluble in water,ethanol,ether and chloroform. |
| Formula Weight | 156.69 |
| Show More |
Safety and Handling
- Tin(II) fluoride
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contain: Stannous fluoride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.