missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) ethoxide

Product Code. 11410510
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410510 1 g 1g
11337977 5 g 5g
11347977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410510 Supplier Thermo Scientific Alfa Aesar Supplier No. 020896.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14791-99-2 | C4H10O2Sn | 208.832 g/mol

Tin(II) ethoxide is used in catalyst synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 14791-99-2
Molecular Formula C4H10O2Sn
Molecular Weight (g/mol) 208.832
MDL Number MFCD00015653
InChI Key XCKWFNSALCEAPW-UHFFFAOYSA-N
Synonym tin ii ethoxide, diethoxytin ii, acmc-20aji5, ethanol, tin 2+ salt 9ci
PubChem CID 71317418
IUPAC Name ethanolate;tin(2+)
SMILES CC[O-].CC[O-].[Sn+2]

Spécification

Melting Point ∼200°C (decomposition)
Linear Formula Sn(OC2H5)2
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water
Formula Weight 208.81
Physical Form Solid/Semi-solid
Chemical Name or Material Tin(II) ethoxide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.