missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) ethoxide

Product Code. 11347977
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347977 25 g 25g
11410510 1 g 1g
11337977 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347977 Supplier Thermo Scientific Alfa Aesar Supplier No. 020896.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14791-99-2 | C4H10O2Sn | 208.832 g/mol

Tin(II) ethoxide is used in catalyst synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14791-99-2
Molecular Formula C4H10O2Sn
Molecular Weight (g/mol) 208.832
MDL Number MFCD00015653
InChI Key XCKWFNSALCEAPW-UHFFFAOYSA-N
Synonym tin ii ethoxide, diethoxytin ii, acmc-20aji5, ethanol, tin 2+ salt 9ci
PubChem CID 71317418
IUPAC Name ethanolate;tin(2+)
SMILES CC[O-].CC[O-].[Sn+2]

Specifications

Melting Point ∼200°C (decomposition)
Linear Formula Sn(OC2H5)2
Quantity 25 g
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water
Formula Weight 208.81
Physical Form Solid/Semi-solid
Chemical Name or Material Tin(II) ethoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.