missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride, ultra dry, 99.9985% (metals basis)

Product Code. 11398577
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398577 25 g 25g
11378577 1 g 1g
11368577 5 g 5g
11388577 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11398577 Supplier Thermo Scientific Alfa Aesar Supplier No. 035723.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7772-99-8 | Cl2Sn | 189.61 g/mol

Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It acts as an antisludging agent and a stabilizer. It can be used to reduce certain metal salts into metals, tin plating and in analytical chemistry to test mercury ions. It is also used as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. It is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Stannous chloride
PubChem CID 24479
ChEBI CHEBI:78067
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C
Melting Point 247°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in water,alkalies,alcohol,methyl ethyl ketone,methyl acetate and acetone.
IUPAC Name dichlorotin
Formula Weight 189.62
Sensitivity Air sensitive; Hygroscopic
Density 3.95
Chemical Name or Material Tin(II) Chloride, Ampouled under argon
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.