missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis)

Product Code. 11327067
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327067 25 g 25g
11337067 5 g 5g
11347067 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327067 Supplier Thermo Scientific Alfa Aesar Supplier No. 010894.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1370709-86-6 | Cl2Sn | 189.61 g/mol

Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1370709-86-6
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Stannous chloride hydrate
PubChem CID 24479
ChEBI CHEBI:78067
SMILES [Cl-].[Cl-].[Sn++]

Specifications

Melting Point 43°C to 46°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula SnCl2·xH2O
UN Number UN3260
Merck Index 14,8783
Solubility Information Soluble in ethanol,acetone,ether and tetrahydrofuran. Insoluble in xylene.
IUPAC Name λ²-tin(2+) dichloride
Formula Weight 189.62 (Anhydrous)
Percent Purity 99.995%
Grade Puratronic
Odor Odorless
Sensitivity Air Sensitive
Density 2.71 g/mL
Assay (metals basis)
Chemical Name or Material Tin(II) chloride hydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tin(II) chloride hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tin(II) chloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.