missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(Ii) Chloride Dihydrate, Reagent grade, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Quantity | 100 g |
| Linear Formula | SnCl2 · 2H2O |
| UN Number | 3260 |
| Solubility Information | Soluble in water |
| Formula Weight | 225.65g/mol |
| Percent Purity | 98% |
| Grade | Reagent |
| Chemical Name or Material | Tin(Ii) Chloride Dihydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction