missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride dihydrate, Reagent Grade

Product Code. 11357227
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
Unit Size:
100g
10g
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357227 10 g 10g
11337227 100 g 100g
11367227 500 g 500g
11347227 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11357227 Supplier Thermo Scientific Alfa Aesar Supplier No. 011536.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10025-69-1 | SnCl2·2H2O

Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-69-1
Molecular Formula SnCl2·2H2O
MDL Number MFCD00149863
Synonym Stannous chloride dihydrate

Specifications

Physical Form Crystalline
Quantity 10 g
Formula Weight 225.65 (189.62 Anhydrous)
Chemical Name or Material Tin(II) chloride dihydrate, Reagent Grade

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.