missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) Chloride Dihydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7073613 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11448962

Brand: Fisher Chemical T/1652/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2Sn·2H2O
MDL Number 149863

Specifications

Boiling Point 652°C
Melting Point 37°C
Color Colorless
Physical Form Solid
Quantity 100 g
Formula Weight 225.63g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.