Learn More
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
£17.40 - £285.00
Chemical Identifiers
CAS | 10025-69-1 |
---|---|
Molecular Formula | Cl2Sn·2H2O |
Molecular Weight (g/mol) | 225.63 |
MDL Number | MFCD00149863 |
Synonym | Stannous chloride dihydrate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10772431
|
Thermo Scientific Acros
196800050 |
5 g |
£17.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10772621
|
Thermo Scientific Acros
196805000 |
500 g |
£73.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10646974
|
Thermo Scientific Acros
196800025 |
2.5 kg |
£285.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-69-1 | |
225.63 | |
Stannous chloride dihydrate |
Cl2Sn·2H2O | |
MFCD00149863 |
Specifications
652.0°C | |
10025-69-1 | |
Adhering Crystalline Powder or Crystals | |
Cl2Sn·2H2O | |
MFCD00149863 | |
15, 8910 | |
Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid | |
225.63 | |
Glass Bottle | |
Tin(II) chloride dihydrate, 97% |
37.0°C to 38.0°C | |
White or Colorless | |
96% min. (Iodimetry) | |
SnCl2·2H2O | |
01,1113; 02,389; 05,631; 06,554; 11,480; 13,298; 15,309; 16,68 | |
Stannous chloride dihydrate | |
225.63 | |
97% | |
2.7100g/mL |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause damage to organs through prolon
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : XP8850000
- Tin(II) chloride dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only