missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Catalog Number 10308253
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7772-99-8
Cl2Sn
189.61
MFCD00011241
AXZWODMDQAVCJE-UHFFFAOYSA-L
Stannous chloride
24479
CHEBI:78067
[Cl-].[Cl-].[Sn++]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7772-99-8
Cl2Sn
189.61
MFCD00011241
AXZWODMDQAVCJE-UHFFFAOYSA-L
Stannous chloride
24479
CHEBI:78067
[Cl-].[Cl-].[Sn++]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 652.0°C
Melting Point 246.0°C
Color White
Physical Form Fine Crystalline Powder or Flakes
Quantity 2.5 kg
Assay Percent Range 97.5 to 102.5% (Iodimetry)
Linear Formula SnCl2
Fieser 01,1113; 02,389; 05,631; 06,554; 11,521; 13,88; 15,138; 16,68; 17,334
Merck Index 15, 8910
Solubility Information Solubility in water: in water: 2700g/L (20°C). Other solubilities: soluble in alkalies, soluble in tartaric acid, alcohol, acetone, ether,, methyl acetate, methyl ethyl ketone and isobutyl, alcohol
IUPAC Name dichlorotin
Formula Weight 189.6
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 3.95
Density 3.9500g/mL
Chemical Name or Material Tin(II) chloride, 98%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Tin(II) chloride
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'