missing translation for 'onlineSavingsMsg'
Learn More

Tin bis(trifluoromethylsulfonyl)imide

Product Code. 15482867
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482867 250 mg 250mg
15462867 1 g 1g
15472867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482867 Supplier Thermo Scientific Alfa Aesar Supplier No. 047142.MD
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4600096-08-6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4600096-08-6
MDL Number MFCD28100488
Synonym Tin bis(trifluoromethanesulfonyl)imide; Tin bis(trifluoromethanesulfonimide)

Specifications

Color White
Linear Formula [(CF3SO2)2N]2Sn
Quantity 250 mg
UN Number UN3261
Sensitivity Hygroscopic
Formula Weight 678.98
Physical Form Solid
Chemical Name or Material Tin bis(trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.