missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solder, Lead-Free, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Solder, Lead-Free, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Aluminium/Gold/Zinc/Cadmium/Iron/Nickel/Bismuth/Indium/Arsenic/Lead/Silver/Antimony/Copper/Tin |
| CAS | 7440-31-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Phosphorus -116 ppm/Aluminium -6 ppm/Gold -5 ppm/Zinc 0.0007 %/Cadmium 0.0015 %/Iron 0.0015 %/Nickel 0.0057 %/Bismuth 0.007 %/Indium 0.007 %/Arsenic 0.01 %/Lead 0.04 %/Silver 0.309 %/Antimony 1.5 %/Copper 4.62 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction