missing translation for 'onlineSavingsMsg'
Learn More

Tin for AAS 10g/L HCl matrix, Fisher Chemical™

Product Code. 12993365 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 12993365

Brand: Fisher Chemical C 155 100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7440-31-5
Physical Form Liquid
Molecular Formula Sn
Concentration 10 g/L ±0.02 g/L
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Type Single-element Concentrated Standard Solution
SMILES [Sn]
IUPAC Name tin
Solvent or Matrix 5% HCl
Molecular Weight (g/mol) 118.71
PubChem CID 5352426
Quantity 100 mL
ChEBI CHEBI:32990
Chemical Name or Material Tin
Show More Show Less
compliance-icons
Product Identifier
  • Single-element Standard Solution for AAS. Tin (Sn) 10000mg/l in HCl 5%
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.