missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin 122, 122Sn, plasma standard solution, Thermo Scientific™, 122Sn 10μg/mL
Click to view available options
Quantity:
50 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Physical Form | Liquid |
MDL Number | MFCD00084602 |
UN Number | UN3264 |
Grade | Specpure™ |
Quantity | 50 mL |
Chemical Name or Material | Tin 122, 122Sn, plasma standard solution, 122Sn 10μg/mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'