missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570423
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%.
Tissue inhibitor of metalloproteinase 3 (TIMP3) is a member of the tissue inhibitor of metalloproteinases gene family. It functions to inhibit the activity of matrix metalloproteinases (MMP) which degrade components of the extracellular matrix. TIMP3 is expressed upon mitogenic stimulation. It binds irreversibly to zinc-dependent MMPs and inactivates them by binding to their catalytic zinc cofactor. Other functions of TIMP3 include nervous system development, tissue regeneration, and visual perception. In humans, the gene encoding TIMP3 is located on chromosome 22.
Specifications
| TIMP3 | |
| Polyclonal | |
| Unconjugated | |
| TIMP3 | |
| HSMRK222; K222; K222TA2; Metalloproteinase inhibitor 3; MIG-5 protein; protein MIG-5; SFD; Sun; TIMP 3; TIMP metallopeptidase inhibitor 3; TIMP metallopeptidase inhibitor 3 (Sorsby fundus dystrophy, pseudoinflammatory); TIMP metallopeptidase inhibitor 3 L homeolog; timp3; TIMP-3; TIMP-3 protein; timp3.L; timp3-A; tissue inhibitor metalloproteinase-3; tissue inhibitor of metalloproteinase 3; tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, pseudoinflammatory); tissue inhibitor of metalloproteinase-3; tissue inhibitor of metalloproteinase-3 precursor; tissue inhibitor of metalloproteinases 3; tissue inhibitor of metalloproteinases-3; XELAEV_18002484mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7078 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P35625 | |
| TIMP3 | |
| synthetic peptide directed towards the middle region of human TIMP3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction