Learn More
Invitrogen™ TIMP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595330
Description
Human TIMP2 shares 98.5% amino acid (aa) sequence identity with both mouse and rat TIMP2. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Spleen Tissue, Rat Testis Tissue, Mouse Brain Tissue, Mouse Thymus Tissue, HELA whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
Specifications
| TIMP2 | |
| Polyclonal | |
| Unconjugated | |
| TIMP2 | |
| Collagenase inhibitor; CSC-21K; D11Bwg1104e; DDC8; etID32613.12; fa96h09; Metalloproteinase inhibitor 2; metalloproteinase inhibitor 2a; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; metalloproteinase-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; TIMP metallopeptidase inhibitor 2a; TIMP2; Timp-2; TIMP-2 protein; timp2a; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase 2 precursor; tissue inhibitor of metalloproteinase 2a; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinase-2 precursor; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2; wu:fa96h09; zgc:136726 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21858, 29543, 7077 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P16035, P25785, P30121 | |
| TIMP2 | |
| E.coli-derived human TIMP2 recombinant protein (Position: C27-P220). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.