missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP2 Mouse, Clone: SPM356, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | TIMP2 |
---|---|
Clone | SPM356 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15950978
|
EnquireBio
7077-MSM1X-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940978
|
EnquireBio
7077-MSM1X-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
TIMP2 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
7077 | |
A synthetic peptide from the N-terminus of human TIMP2 protein (aa56-66) (DSGNDIYGNPI) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
21kDa |
SPM356 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_003246 | |
TIMP2 | |
IgG2a κ | |
Protein A/G Purified | |
Cytoplasm and Cell Surface | |
HeLa cells. Normal Placenta. Breast, Colon, Endometrial, Prostate or Ovarian Carcinoma. |