missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMP1 Monoclonal Antibody (C8), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £361.00
Specifications
| Antigen | TIMP1 |
|---|---|
| Clone | C8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17909252
![]() |
Invitrogen™
MA551017 |
200 μL |
£361.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17920413
|
Invitrogen™
MA551018 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.Specifications
| TIMP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| CLGI; collagenase inhibitor; Collagenase inhibitor 16C8 fibroblast; EG-1; Embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; Timp; TIMP 1; TIMP metallopeptidase inhibitor 1; TIMP1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metal proteinases; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1 | |
| TIMP1 | |
| IgG1 κ | |
| Protein A/G | |
| TIMP1 |
| C8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat | |
| P12032, P30120 | |
| 116510, 21857 | |
| Recombinant Rat TIMP1 protein, Cys24-Ala217 (Accession #P30120). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title