missing translation for 'onlineSavingsMsg'
Learn More

TIMP-4 Antibody (SB30c), DyLight 755, Novus Biologicals™

Product Code. 18417505 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417505 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417505 Supplier Novus Biologicals Supplier No. NBP128619IR

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TIMP-4 Monoclonal antibody specifically detects TIMP-4 in Human, Mouse samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP-4
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SB30c
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4
Host Species Mouse
Immunogen Recombinant human TIMP-4 (a.a. 35-224)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7079
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.