missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-4 Antibody (SB30c), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28619AF532
This item is not returnable.
View return policy
Description
TIMP-4 Monoclonal specifically detects TIMP-4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| TIMP-4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TIMP4 | |
| Recombinant human TIMP-4 (a.a. 35-224) | |
| 0.1 mL | |
| Primary | |
| Human/Mouse/Rat TIMP-4 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| SB30c | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4 | |
| Mouse | |
| Protein A or G purified | |
| Angiogenesis, Cancer, Cellular Markers, Hypoxia | |
| 7079.0 | |
| Human, Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction