missing translation for 'onlineSavingsMsg'
Learn More

TIMP-4 Antibody (866106R) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30734082 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734082 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734082 Supplier Novus Biologicals Supplier No. FAB7667I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

TIMP-4 Monoclonal antibody specifically detects TIMP-4 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP-4
Applications ELISA
Classification Monoclonal
Clone 866106R
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TIMP-4, Met1-Pro224, Accession # Q9JHB3
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7079
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.